Would you like to log in again?
Human Recombinant IL-7 (from E. coli)
Supplier: PeproTech
200-07-10UG 200-07-100UG 200-07-2UG 200-07-500UG 200-07-250UG 200-07-50UG
10779-680EA 2793.41 USD InStock
10779-680 10779-672 10779-682 10779-676 10779-674 10779-678
Human Recombinant IL-7 (from E. coli)
Proteins and Peptides
IL-7 is a hematopoietic growth factor that primarily affects early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors, such as ConA and IL-2. Human and murine IL-7 are cross-species reactive. Recombinant Human IL-7 is a 17.4 kDa protein containing 153 amino acid residues.
  • Carrier free
  • Verified by N-terminal and Mass Spectrometry analyses (when applicable)
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty