Would you like to log in again?
Sklarlite XD™ Thumb Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
23-2819
10195-352EA 93.58 USD InStock
10195-352
Sklarlite XD™ Thumb Tissue Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures.
They have spring tension shanks that determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are toothed to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference.
Sklarlite XD™ instruments are lighter and delicate than standard instruments, which allow the surgeon high precision with the least amount of fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty