Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 35%, Technical Grade

Supplier: Columbus Chemical Industries, Inc
Danger

268500-80401
71001-588CS 223.93 USD InStock
71001-588
Hydrogen peroxide 35%, Technical Grade
Hydrogen peroxide


Formula: H₂O₂
MW: 34.01 g/mol
Melting Pt: –33 °C
Density: 1.13 g/cm³
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Assay (H2O2) 35.1 - 35.9%
Color (APHA) 22 max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty