Would you like to log in again?
Pean Gastro-Intestinal Forceps, OR Grade, Sklar
Supplier: Sklar
36-2890 36-2812 36-2990
10195-760EA 272.9 USD InStock
10195-760 10195-758 10195-756
Pean Gastro-Intestinal Forceps, OR Grade, Sklar
Forceps
Sklar's® Rochester-Pean Forceps are designed with longer serrated jaws for clamping large vessels
  • Ratcheted finger rings prevents slipping while clamping tissue
  • Available straight or curved
  • Variety of lengths eithe longitudinal or horizontal serrations
Censitrac Ready products are industry standard and constructed of stainless steel.
These grasping forceps appropriate for temporary occlusion of the intestine, or approximation of the intestine edges during anastomosis procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty