Would you like to log in again?
Streptavidin Agarose, Novagen®, MilliporeSigma
Supplier: MilliporeSigma
Novagen®
69203-3 69203-3
EM69203-3EA 569.53 USD InStock
EM69203-3 CA80030-966
Streptavidin Agarose, Novagen®, MilliporeSigma
Protein Purification Protein Purification Resins
Streptavidin agarose for column or batch isolation of biotinylated molecules.
  • Cross-linked agarose covalently coupled with pure streptavidin
  • Sutiable for use in column and batch formats
  • Batch isolation of biotinylated molecules
It consists of cross-linked agarose covalently coupled with pure streptavidin under controlled conditions
Provided as a 50% slurry in storage buffer; 5 mL settled resin in 10 mL total volume. 100 mM sodium phosphate pH 7.5, 0.02% sodium azide is a storage buffer and +2°C to +8°C is ideal temperature range for storage of agarose.
This stable link minimizes the leaching of the streptavidin while maintaining full binding activity. This matrix is suitable for use in column and batch formats for any application that requires high biotin binding capacity and low non-specific binding. The resin has no detectable protease, DNase or RNase contamination.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty