Would you like to log in again?
Doyen Forceps, OR Grade, Sklar
Supplier: Sklar
36-3690 36-3590
10195-772EA 489.4 USD InStock
10195-772 10195-770
Doyen Forceps, OR Grade, Sklar
Forceps
Doyen Forceps are a lightweight, non-crushing clamp used to grasp large organs such as lungs, or intestines
Censitrac Ready products are industry standard and constructed of stainless steel.
They are available straight or curved, are slightly malleable, and feature longitudinal serrations, with long jaws and shorter shanks.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty