Would you like to log in again?
Allis-Adair Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
36-1765
10195-728EA 205.93 USD InStock
10195-728
Allis-Adair Tissue Forceps, OR Grade, Sklar
Forceps
Allis-Adair Forceps are a ratcheted tissue grasping instrument used to grip and hold wider sections of tissue
This product provides a secure hold on larger sections of tissue, that need to be held out of the surgery site.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty