Would you like to log in again?
Econo™ Mayo Dissecting Scissors, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-308 21-321 21-325 21-312
89023-150PK 83.26 USD InStock
89023-150 89023-144 89023-146 89023-154
Econo™ Mayo Dissecting Scissors, Floor Grade, Sklar
Scissors
Econo™ Mayo Dissecting scissors are a versatile instrument that can be used in multiple procedures and multiple settings.
The curved instrument is used for cutting or dissecting dense or deep tissue, and the straight version is used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Econo™ instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty