Would you like to log in again?
Pine-Sol® Multi-Surface Cleaner
Supplier: Essendant
Pine-Sol®
CLO35418CT
102877-766CS 109.54 USD InStock
102877-766
Pine-Sol® Multi-Surface Cleaner
Cleaning Agents Detergents and Surface Cleansers
Clean, deodorize and disinfect with the power of Pine-Sol
Applicable materials include Ceramics; Enamel; Fiberglass; Finished/Sealed Wood; Laminates; Metal; Plastic; Sealed/Engineered Stone; Vinyl. Dirt types include Bacteria; Dust; Fungus; Germs; Grease; Grime; Mildew; Mold; Viruses; Water-Based Soil.
Note: Do not use on marble, aluminum or unfinished, unsealed, unpainted, waxed, oiled or worn wood.
Cuts through tough grease and grime fast while leaving behind a clean scent. No sticky residue. Use on a wide variety of tough jobs and surfaces including floors, counters and sinks. Perfect for use in offices, day care centers, restaurants, schools and other commercial facilities. Liquid. Pine scent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty