Would you like to log in again?
Endust® Antistatic Cleaner for Electronics, Endust
Supplier: Endust
Endust®
END096000
500044-764EA 5.23 USD InStock
500044-764
Endust® Antistatic Cleaner for Electronics, Endust
Cleaning Agents Detergents and Surface Cleansers
Versatile spray cleans all monitor-screen types as well as just about every surface in your office.
  • No-run formula stays where you spray it
  • Removes dust and fingerprints while helping to prevent static-charge build-up
  • Dries streak free
  • For best results, use with XL microfiber wipes
Applicable materials include Ceramics; Fiberglass; Glass; Laminates; Metal; Plastic. Dirt types include Debris; Dust; Grease; Fingerprints; Soil; Smudges. Liquid. Aerosol.
Alcohol-free to safely clean all of your valuable electronics: tablets, televisions, computers, smartphones and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty