Would you like to log in again?
Surgi-OR™ Adson Dressing Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-776 95-775
89023-714EA 35.53 USD InStock
89023-714 89023-716
Surgi-OR™ Adson Dressing Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Adson Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow.
Surgi-OR™ mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty