Would you like to log in again?
Surgi-OR™ Rankin Kelly Hemostatic Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-465 95-469
89023-632EA 50.38 USD InStock
89023-632 89023-630
Surgi-OR™ Rankin Kelly Hemostatic Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Rankin Kelly Hemostatic Forceps are used for clamping large blood vessels, or manipulating heavy tissue
They are available curved or straight with a ratcheted finger ring handle. Similar to Kelly forceps, the jaws of Rankin Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Rankin forceps are longer and lighter than Kelly forceps.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
They may also be used for soft tissue dissection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty