Would you like to log in again?
Lawrence Forceps, OR Grade, Sklar
Supplier: Sklar
55-2685
10197-570EA 315.12 USD InStock
10197-570
Lawrence Forceps, OR Grade, Sklar
Forceps
Lawrence Forceps are a ratcheted, finger ring instrument used for controlling blood flow is vessels, often in cardiovascular and thoracic procedures.
The long shanks allow this instrument to be used in difficult to reach areas, while the curved, serrated jaws can clamp obscured vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty