Would you like to log in again?
Kraff-Utrata Capsulorhexis Forceps, OR Grade, Sklar
Supplier: Sklar
66-5351
10198-024EA 882.52 USD InStock
10198-024
Kraff-Utrata Capsulorhexis Forceps, OR Grade, Sklar
Forceps
Kraff-Utrata Capsulorhexis Forceps are an ophthalmic instrument used during cataract procedures
These forceps feature cystotome tips, which are sharp and needle like, and used to puncture and create an initial cut in the membrane. The jaws of the forceps are then used to tear, in a C-shaped motion, and lift the membrane surrounding the clouded lens (the cataract) so it can be removed. The lens is replaced with a plastic, prescription lens. This product is has angled jaws that are 12-mm long with .33-mm fine tips.
In order to remove a clouded lens (a cataract), a thin membrane (the capsule), that surrounds the lens must be incised.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty