Would you like to log in again?
Econo™ Pederson Vaginal Speculum, Sklar
Supplier: Sklar
Econo™
21-348 21-362
89023-168PK 512.12 USD InStock
89023-168 89023-174
Econo™ Pederson Vaginal Speculum, Sklar
Specula
Econo™ Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix.
  • Industry standard
  • Ready to use
  • Disposable and latex-free
These are a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are narrower than the blades of the Graves Speculum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty