Would you like to log in again?
Friedman Rongeur, OR Grade, Sklar
Supplier: Sklar
93-1603
94002-760EA 415.21 USD InStock
94002-760
Friedman Rongeur, OR Grade, Sklar
Bone Cutters and Rongeurs
Sklar's® Friedman Bone Rongeur is a double spring, plier handle instrument used for debulking and removing pieces of bone.
  • Industry standard
This rongeur has delicate jaws for use with delicate ear, nasal, and orbital bones. The slight curvature of the jaws makes this instrument useful in these obscure areas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty