Would you like to log in again?
Sklarlite™ Rochester-Pean Forceps, OR Grade, Sklar
Supplier: Sklar
23-2532 23-2542
10195-332EA 233.26 USD InStock
10195-332 10195-160
Sklarlite™ Rochester-Pean Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures.
They are available curved or straight. Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty