Would you like to log in again?
Hartmann Nasal Forceps, OR Grade, Sklar
Supplier: Sklar
70-4160
10198-528EA 373.22 USD InStock
10198-528
Hartmann Nasal Forceps, OR Grade, Sklar
Forceps
Hartmann Nasal Forceps are used in rhinology and rhinoplasty.
Hartmann's Nasal Forceps have slightly shorter shanks and are not as angled as Alligator Forceps. This product is angled at the box lock with straight, serrated, blunt tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty