Would you like to log in again?
Sawtell Tonsil Forceps, OR Grade, Sklar
Supplier: Sklar
74-3175
10198-614EA 197.23 USD InStock
10198-614
Sawtell Tonsil Forceps, OR Grade, Sklar
Forceps
Sklar's® Sawtell Tonsil Forceps are ratcheted, finger ring forceps used primarily for clamping tissue and holding sponges during tonsillectomy procedures.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty