Would you like to log in again?
Gradle Cilia Forceps, OR Grade, Sklar
Supplier: Sklar
66-1745
10194-390EA 381.81 USD InStock
10194-390
Gradle Cilia Forceps, OR Grade, Sklar
Forceps
Gradle Cilia Forceps are thumb forceps used to grasp and hold cilia, or eyelashes.
These forceps have a double leaf spring operating mechanism, different from other cilia forceps. The jaws are very short, beginning after a screw lock, and curve slightly inward. They have smooth, blunt, squared tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty