Would you like to log in again?
Potts-Smith Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
52-3370
10194-466EA 123.43 USD InStock
10194-466
Potts-Smith Tissue Forceps, OR Grade, Sklar
Forceps
Potts-Smith Forceps are medium weight thumb forceps used for grasping fine or delicate tissue.
They securely grasp the tissue, but also have a stop peg to prevent an overly harsh grasp that may crush the tissue. These forceps are used on fine or single layer tissues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty