Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Merit™ Allis Tissue Forceps, Physician Grade, Sklar
Supplier: Sklar
Merit™
97-930 97-938 97-934
10184-936EA 30.99 USD InStock
10184-936 10194-484 10184-938
Merit™ Allis Tissue Forceps, Physician Grade, Sklar
Forceps
Merit™ Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue.
They are used with tonsil, vaginal, breast, and thyroid tissue.
Merit™ instruments are office-grade line of products used for routine and in-office procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty