Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ballenger Sponge Forceps, OR Grade, Sklar®
Supplier: Sklar
74-1470 74-1370 74-1570 74-1670
10198-606EA 157.35 USD InStock
10198-606 10192-476 10198-602 10198-604
Ballenger Sponge Forceps, OR Grade, Sklar®
Forceps
Ballenger Sponge Forceps are a ratcheted and finger ring instrument used to grasp tissue or hold sponges
Sponge Forceps may also be known as sponge sticks. The instrument is used to hold gauze or cotton, that is wrapped around the tips and the ratcheted is locked to secure a firm grip. This creates a sponge stick that can be used in any type of procedure to absorb blood and excess fluids from the surgical site and surrounding areas.
They may be known as Foerster Sponge Forceps, however the shorter length instrument is often referred to as Ballenger.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty