Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Foerster Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
66-6038
10198-030EA 490.48 USD InStock
10198-030
Foerster Tissue Forceps, OR Grade, Sklar
Forceps
Foerster Tissue Forceps are thumb forceps used for handing fine, delicate tissue during ophthalmic procedures.
Tissue Forceps, unlike Dressing Forceps, are toothed so they can grip tissue, which may be slippery. The handle features a fenestrated octagonal thumb/finger grip for increased precision, control, and comfort.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty