Would you like to log in again?
Jones Forceps, OR Grade, Sklar
Supplier: Sklar
17-1255 17-1355
76192-104EA 173.53 USD InStock
76192-104 10194-744
Jones Forceps, OR Grade, Sklar
Forceps
Jones Forceps, also known as Spencer-Wells forceps, are ratcheted, finger ring hemostatic forceps.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Jones Forceps are ratcheted, finger ring hemostatic forceps used to control blood flow in vessels. They are similar to Lahey forceps, having serrations the entire length of the jaw. However, the jaw is slightly shorter than that of the Lahey.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty