Would you like to log in again?
Marcuse Tubing Forceps, OR Grade, Sklar
Supplier: Sklar
17-3980 17-3955
10194-814EA 203.12 USD InStock
10194-814 10194-818
Marcuse Tubing Forceps, OR Grade, Sklar
Forceps
Sklar's® Marcuse Tubing Forceps is a ratcheted, finger ring instrument used to block the flow of fluids from large chest tubes or tubing from bypass machines
The beveled, smooth jaws provide a tight grip on tubing, while the ratcheted locking mechanism can hold that grip in place.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty