Would you like to log in again?
Sklar Black™ Thumb Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
90-1022 90-1020
10199-048EA 254.56 USD InStock
10199-048 10199-046
Sklar Black™ Thumb Tissue Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in laser procedures.
These instruments have spring tension shanks, that determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Ordering information: Available in several lengths to accommodate specific procedures, tissue types, and user preference.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty