Would you like to log in again?
Chandler Iris Forceps, OR Grade, Sklar
Supplier: Sklar
66-3927
10197-500EA 236.26 USD InStock
10197-500
Chandler Iris Forceps, OR Grade, Sklar
Forceps
Chandler Iris Forceps are opthalmologic thumb forceps used for handling the delicate tissue of the iris.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty