Would you like to log in again?
Rankin Forceps, OR Grade, Sklar
Supplier: Sklar
17-1262 17-1362
10193-826EA 83.96 USD InStock
10193-826 10192-266
Rankin Forceps, OR Grade, Sklar
Forceps
Sklar® Kelly-Rankin Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue.
Sklar Kelly-Rankin Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue
Censitrac Ready, Rankin Forceps are industry standard and constructed of stainless steel.
The jaws are 1/3 the length of the shanks, and serrations are 1/2 the length of the jaws. Kelly forceps that have an overall length of 6.25" are referred to as Kelly-Rankin forceps.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty