Would you like to log in again?
DeBakey-Derra Atraumatic Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
52-6652 52-6650
10196-244EA 709.39 USD InStock
10196-244 10196-246
DeBakey-Derra Atraumatic Forceps, OR Grade, Sklar
Forceps
DeBakey-Derra Atraumatic Forceps are a ratcheted, finger ring instrument used for anastomosis in cardiovascular procedures
It is a partially occluding clamp, that is used to control blood flow from the carotid artery, aorta, and inferior vena cava. The atraumatic teeth provide a secure and delicate grip on vessels, causing little to no damage to the vessel walls. This product has slightly curved shanks with a jaw type, that continues past the lock box into the jaws.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty