Would you like to log in again?
Cooley Vascular Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
52-6690 52-6694 52-6696
10196-268EA 340.24 USD InStock
10196-268 10196-270 10196-272
Cooley Vascular Tissue Forceps, OR Grade, Sklar
Forceps
Cooley Vascular Tissue Forceps are a thumb forceps used in cardiothoracic procedures to grasp delicate tissue.
The Cooley style teeth cause little to no damage, and the narrow tips can be placed in narrow areas. The thumb grip is wider than the tips, so the physician will have adequate control.
OR grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. These instruments have been designed to provide comfort and precision, while ensuring the durability and strength, that has been provided for over 100 years.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty