Would you like to log in again?
Selman Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
52-4260 52-4310 52-4280
10196-444EA 376.82 USD InStock
10196-444 10196-446 10196-448
Selman Tissue Forceps, OR Grade, Sklar
Forceps
Selman Tissue Forceps are thumb forceps used in cardiac and vascular procedures.
The fine, meshing teeth of these non-slip forceps hold tissue and vessels without damaging them.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty