Would you like to log in again?
Gradle Stitch Scissors, OR Grade, Sklar
Supplier: Sklar
22-2957
89025-344EA 212.07 USD InStock
89025-344
Gradle Stitch Scissors, OR Grade, Sklar
Scissors
Gradle Stitch Scissors are used for suture removal.
These scissors have short blades with sharp, fine tips that are used to snip the suture rather than lifting the suture slightly first, as with Spencer or Littauer stitch scissors.
Gradle Stitch Scissors should not be used for cutting tissue, but may be useful for delicate or micro-dissection. This product is slightly curved with smooth, sharp/sharp tips and a length of 3-3/4 inches. It also features Ribbon Style Finger Ring handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty