Would you like to log in again?
Undermining Kaye Scissors, OR Grade, Sklar
Supplier: Sklar
47-1084
10196-828EA 163.11 USD InStock
10196-828
Undermining Kaye Scissors, OR Grade, Sklar
Scissors
Sklar's® Kaye undermining ribbon handled scissors are used in the platic and general surgery fields, common in face lift procedures
Undermining Kaye Scissors and Censitrac Ready products are industry standard and constructed of stainless steel.
They are a lighter pattern than the Gorney-Freemans.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty