Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
ADI-SAB-200-J
10049-452EA 195.09 USD InStock
10049-452
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Immunoglobulin G (IgG) is an immunoglobulin monomer consisting of two (gamma) heavy chains and two light chains. Each IgG molecule contains two antigen binding domains and a single effector (Fc) domain. Assay Designs offers isotyping kits and antibodies in a variety of formats for the detection of human, mouse, rabbit, rat, sheep, and goat IgG.

Species reactivity: Rat

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty