Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amyl acetate ACS

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Acetic acid amyl estern-Pentyl acetatePentyl acetateAcetic acid pentyl ester

10800 10810
100503-658EA 427.27 USD InStock
100503-658 100503-656
Amyl acetate ACS
Amyl acetate


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149.25 °C (301 °F)
Melting Pt: –70.8 °C (–95 °F)
Density: 0.8752 g/cm³ (7.304 lbs/gal.)
Flash Pt: 16…25 °C (61…77 °F)
MDL Number: MFCD00009500
CAS Number: 628-63-7
UN: 1104
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/77/28984777.jpg
Order Now
Specification Test Results
Acidity (as CH₃COOH) 0.01%
Non-volatile Matters 0.01%
Specific Gravity at 25 °C 0.870 - 0.875
Water (W/W) 0.2 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty