Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.95%, powder ACS

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

11460
100503-696EA 75.49 USD InStock
100503-696
Boric acid ≥99.95%, powder ACS
ortho-Boric acid
A buffer component in Tris-EDTA-Borate buffer. Also used in silver-methenamine buffer according to Swift, J.A. (1968).

The Electron histochemistry of cystine-containing proteins in thin transverse sections of human hair (J.R. Microsc. Soc. 88,449).


Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 300 °C (572 °F)
Melting Pt: 160 °C (320 °F)
Density: 1.44 g/cm³ (12.017 lbs/gal.)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty