Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oil of cedar wood clearing reagent

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Cedar Leaf OilCedar wood oilCedar Oil

12420
100496-084EA 110.9 USD InStock
100496-084
Oil of cedar wood clearing reagent
Oil of cedar wood
Can be used for clearing.


Density: 0.947 g/cm³ (7.903 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00146327
CAS Number: 8000-27-9
Order Now
Specification Test Results
Refractive Index at 20 °C 1.504 ±0.002

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty