Would you like to log in again?
Chemkit for Standard Preparations of Biological Specimens, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
14750
100491-320KT 1454.5 USD InStock
100491-320
Chemkit for Standard Preparations of Biological Specimens, Electron Microscopy Science
Stains Microscopy and staining reagents
Chemkit is arranged to enable a laboratory to perform standard preparations of biological specimens.
This package enables a laboratory to perform standard preparations of biological specimens. Includes fixatives-osmium tetroxide crystal, osmium tetroxide 4% solution, glutaraldehyde EM grade 50%, glutaraldehyde EM grade 8%, and paraformaldehyde Prill.
Buffers include sodium phosphate monobasic, sodium phosphate dibasic, sodium hydroxide pellets, and sodium cacodylate trihydrate. Dehydrants include propylene oxide and acetone.
For embedding media the 812 kit and low viscosity kit (Dr. Spurr) are included. Included stains are toluidine blue, borax tetra, sodium borate, uranyl acetate, lead citrate, and epoxy tissue stain.
Delivery information: Kit includes osmium tetroxide crystal (5×1 g), osmium tetroxide 4% solution (10×2 ml), glutaraldehyde EM Grade 50% (10×10 ml), glutaraldehyde EM grade 8% (10×10 ml), paraformaldehyde, prill (1 kg), 16% paraformaldehyde (formaldehyde) aqueous solution (10×10 ml) fixations. Buffers like sodium phosphate monobasic (500 g), sodium phosphate dibasic (500 g), sodium hydroxide pellets (500 g), sodium cacodylate trihydrate (500 g). Dehydrants, propylene oxide (4×450 ml), acetone (4×1 qt) and embedding media includes EMbed-812 kit, low viscosity kit, toluidine blue (50 g), borax tetra, sodium borate (500 g), uranyl acetate (25 g), lead citrate (2×25 g) and epoxy tissue stain (795.00).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty