Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maleic acid, crystalline ACS, repurified

Supplier: Electron Microscopy Sciences
Warning

18150
100504-094EA 77.29 USD InStock
100504-094
Maleic acid, crystalline ACS, repurified
Maleic acid


Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 160 °C (320 °F)
Density: 1.59 g/cm³ (13.269 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
UN: 1759
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/48/28984848.jpg
Order Now
Specification Test Results
Assay 99.5%
Chloride 0.01%
Fumaric Acid 0.15%
Insoluble Matters 0.005%
Sulfate 0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty