Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methacrylic acid glacial ≥98% stabilized

Supplier: Electron Microscopy Sciences
Danger
Synonyms: 2-Methylpropenoic acid

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 0.25%

18500
100504-952EA 66.17 USD InStock
100504-952
Methacrylic acid glacial ≥98% stabilized
Methacrylic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 161 °C (322 °F)
Melting Pt: 14 °C (57 °F)
Density: 1.0153 g/cm³ (8.473 lbs/gal.)
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
UN: 2531
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/49/28984849.jpg
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty