Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl green zinc chloride salt, certified (C.I. 42590)

Supplier: Electron Microscopy Sciences
Danger
Synonyms: CI 42590Methyl green zinc chloride double saltMethyl GreenEthyl Green

18700
100504-108EA 119.05 USD InStock
100504-108
Methyl green zinc chloride salt, certified (C.I. 42590)
Methyl Green zinc chloride salt
Used as a counterstain in Almann's aniline acid fuchsin technique for mitochrondria; used in Kurnick's Methyl Green-Pyonin stain for nuclei and cystoplasmic granules of liver cells. Marker for DNA at acid and neutral conditions (J.B. Boyd, H.R. Mitchell, (1965) Anal. Biochem., 13, 28). Used for epoxy-embedded thick sections (Sievers, J. (1971)).

Solubility : 8.0% water; 3.0% Alcohol; 1.8% Cellosolve; 3.5% Glycol; 0.0% Xylene


Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24
Melting Pt: 233 °C (451 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
UN: 2923
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/52/28984852.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty