Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl violet 2B, certified (C.I. 42535)

Supplier: Electron Microscopy Sciences
Synonyms: Basic Violet 14-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride

18850
100504-112EA 82.59 USD InStock
100504-112
Methyl violet 2B, certified (C.I. 42535)
Methyl Violet 2B
Used with methyl green to stain epoxy-embedded sections.


Formula: C₂₄H₂₈ClN₃
MW: 393.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Dye Content Min. 75%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty