Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nile Blue A, certified (C.I. 51180)

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Basic Blue 12Nile blue A sulfateNile blue sulphateNile blue A sulphateNile blue sulfate

19020
100504-120EA 35.49 USD InStock
100504-120
Nile Blue A, certified (C.I. 51180)
Nile blue A
Differentiates melamins and lipofuscins; Used in fat staining.

Lillie, Stain Tech., 31,151 (1956); J. Histochem., 4,377; Z. Anal. Chem. 260, 290 (1972).
For staining bone sections. Cytologia, 11, 261 (1940).


Formula: C₄₀H₄₀N₆O₆S
MW: 732.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064529
CAS Number: 3625-57-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/64/28984864.jpg
Order Now
Specification Test Results
Dye Content Approx. 65%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty