Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecamolybdophosphoric acid hydrate, crystals ACS

Supplier: Electron Microscopy Sciences
Danger
Synonyms: 12-Molybdophosphoric acid hydrateMolybdophosphoric acid hydratePhosphomolybdic acid hydrate

19400
100504-192EA 120.64 USD InStock
100504-192
Dodecamolybdophosphoric acid hydrate, crystals ACS
Dodecamolybdophosphoric acid hydrate


Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Density: 1.62 g/cm³ (13.519 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
UN: 3084
ADR: 8,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty