Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Picric acid, crystals ACS

Supplier: Electron Microscopy Sciences
Danger

19550
100504-196EA 184.74 USD InStock
100504-196
Picric acid, crystals ACS
Picric acid


Formula: C₆H₃N₃O₇
MW: 229.11 g/mol
Boiling Pt: 301 °C (574 °F)
Density: 1.767 g/cm³ (14.746 lbs/gal.)
Flash Pt: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007102
CAS Number: 88-89-1
UN: 1344
ADR: 4.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/77/28984877.jpg
Order Now
Specification Test Results
Insoluble in Benzene 0.1%
Insoluble in Resinous Matter 0.01%
Sulfate 0.01%
Water Min. 30%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty