Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloromethane, Plastisolve

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Methane dichlorideMethylene chloride

19610 19600
100504-200EA 67.81 USD InStock
100504-200 100504-202
Dichloromethane, Plastisolve
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39 °C (102 °F)
Melting Pt: –95.1 °C (–139 °F)
Density: 2.93 g/cm³ (24.451 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty