Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexacyanoferrate(III) ACS

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Potassium ferricyanide(III)Potassium ferricyanideTripotassium hexacyanoferrate

20150
100504-230EA 99.35 USD InStock
100504-230
Potassium hexacyanoferrate(III) ACS
Potassium hexacyanoferrate (III)


Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.89 g/cm³ (15.772 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
Order Now
Specification Test Results
Chloride 0.01%
Ferro compounds (Fe(Cn)₄ 0.05%
Insoluble matters 0.005%
Sulfate 0.01%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty