Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safranine O, certified (C.I. 50240)

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

20800
100504-260EA 72.13 USD InStock
100504-260
Safranine O, certified (C.I. 50240)
Safranine O
(Safranin T, Y or A; Basis Red 2; Cotton Red; Gossypimine)
C20H19N4Cl F.W. 350.85
Solubility: 4.5% Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% XylenePrussian Blue method for hemosiderin. Weigert's iron hematoxylin with methachromic dyes which stain nuclei and granules. A alkaline phosphate stain. Flemming's method for staining chromatin and nuclear elements.

Certified by the Biological Stain Commission All Dyes/Stains are for research use only. Not meant for drug, food household use.

Solubility : Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% Xylene


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Density: 1 g/cm³ (8,345 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty