Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutaraldehyde

Supplier: Electron Microscopy Sciences
Synonyms: 1,5-PentanedialGlutardialdehydeGlutaral

16216 16316 16314 16214
100503-996EA 121.69 USD InStock
100503-996 100503-994 100503-984 100503-982
Glutaraldehyde
Glutaraldehyde
25% and 50% EM Grade Aqueous Glutaraldehyde in packaged in serum vials. Packaging allows puncture special non-reactive seal with a hypodermic syringe and draw out exact quantity of Glutaraldehyde needed without opening container. Assures freshness and purity of Glutaraldehyde at all times.


Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00007025
CAS Number: 111-30-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty